missing translation for 'onlineSavingsMsg'
Learn More

4-Butyl-2-methylaniline, Tech., Thermo Scientific™

Product Code. p-2701320
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10477242

Brand: Thermo Scientific Maybridge SB01607DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 72072-16-3
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00190660
InChI Key JTXOXRXZCAMPHL-UHFFFAOYSA-N
PubChem CID 577772
IUPAC Name 4-butyl-2-methylaniline
SMILES CCCCC1=CC(=C(C=C1)N)C

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Synonym benzenamine,4-butyl-2-methyl, 2-methyl-4-butylaniline, 4-butyl-2-methyl-aniline, 4-n-butyl-2-methylaniline, 4-butyl-2-methylphenylamine, #, benzenamine, 4-butyl-2-methyl
Formula Weight 163.26
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-Butyl-2-methylaniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.