missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoveratrole, 97%

Product Code. 11429033
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429033 500 g 500g
11409033 25 g 25g
11419033 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429033 Supplier Thermo Scientific Alfa Aesar Supplier No. A11133.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2859-78-1 | C8H9BrO2 | 217.062 g/mol

4-Bromo-1,2-dimethoxybenzene is a metabolite of bromobenzene, with a catechol moiety in the substructure. It also has use as a redox shuttle additive, essentially a component in lithium batteries that consumes excess current during overcharge. And also used in the synthesis of the isoquinoline alkaloid (-)-mesembrine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-1,2-dimethoxybenzene is a metabolite of bromobenzene, with a catechol moiety in the substructure. It also has use as a redox shuttle additive, essentially a component in lithium batteries that consumes excess current during overcharge. And also used in the synthesis of the isoquinoline alkaloid (-)-mesembrine.

Solubility
Soluble in benzene, ether, toluene.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2859-78-1
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.062
MDL Number MFCD00008381
InChI Key KBTMGSMZIKLAHN-UHFFFAOYSA-N
Synonym 4-bromoveratrole, 1-bromo-3,4-dimethoxybenzene, 3,4-dimethoxybromobenzene, p-bromoveratrole, benzene, 4-bromo-1,2-dimethoxy, 3,4-dimethoxyphenyl bromide, 4-bromo-1,2-dimethoxy-benzene, 1,2-dimethoxy-4-bromobenzene, 4-bromocatechol dimethyl ether, 4-bromoveratrol
PubChem CID 76114
IUPAC Name 4-bromo-1,2-dimethoxybenzene
SMILES COC1=C(C=C(C=C1)Br)OC

Specifications

Density 1.51
Boiling Point 255°C to 256°C
Flash Point 109°C (228°F)
Refractive Index 1.573
Quantity 500 g
Beilstein 2046884
Solubility Information Soluble in benzene,ether,toluene.
Formula Weight 217.06
Percent Purity 97%
Chemical Name or Material 4-Bromoveratrole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.