missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromovalerophenone, 98%, Thermo Scientific™

Product Code. 11431557
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431557 100 g 100g
11411557 5 g 5g
11421557 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431557 Supplier Thermo Scientific Alfa Aesar Supplier No. B24979.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7295-44-5
Molecular Formula C11H13BrO
Molecular Weight (g/mol) 241.128
MDL Number MFCD00000107
InChI Key STYJKBMRWQQJIS-UHFFFAOYSA-N
Synonym 4'-bromovalerophenone, 1-4-bromophenyl pentan-1-one, p-bromophenyl butyl ketone, p-bromovalerophenone, 1-pentanone, 1-4-bromophenyl, 4-bromovalerophenone, 1-bromo-4-pentanoylbenzene, 1-4-bromophenyl-1-pentanone, acmc-20ammf, 4;-bromovalerophenone
PubChem CID 81717
IUPAC Name 1-(4-bromophenyl)pentan-1-one
SMILES CCCCC(=O)C1=CC=C(C=C1)Br

Specifications

Melting Point 34°C to 36°C
Boiling Point 168°C to 169°C (20 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Formula Weight 241.14
Percent Purity 98%
Chemical Name or Material 4'-Bromovalerophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.