missing translation for 'onlineSavingsMsg'
Learn More

4-Bromotriphenylamine, 99%, Thermo Scientific Chemicals

Catalog Number 15416798
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
36809-26-4
C18H14BrN
324.221
MFCD01851266
SQTLUXJWUCHKMT-UHFFFAOYSA-N
4-bromotriphenylamine, 4-bromo-phenyl-diphenyl-amine, 4-bromophenyl diphenylamine, 4-bromo triphenylamine, benzenamine, 4-bromo-n,n-diphenyl, n,n-diphenyl-4-bromoaniline, rarechem fh 1g 0a02, n-4-bromophenyl-n-phenylbenzenamine, 4-diphenylamino-1-bromobenzene
642838
4-bromo-N,N-diphenylaniline
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)Br
This item is not returnable. View return policy
36809-26-4
C18H14BrN
324.221
MFCD01851266
SQTLUXJWUCHKMT-UHFFFAOYSA-N
4-bromotriphenylamine, 4-bromo-phenyl-diphenyl-amine, 4-bromophenyl diphenylamine, 4-bromo triphenylamine, benzenamine, 4-bromo-n,n-diphenyl, n,n-diphenyl-4-bromoaniline, rarechem fh 1g 0a02, n-4-bromophenyl-n-phenylbenzenamine, 4-diphenylamino-1-bromobenzene
642838
4-bromo-N,N-diphenylaniline
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 108°C to 112°C
Quantity 5 g
Formula Weight 324.21
Percent Purity 99%
Chemical Name or Material 4-Bromotriphenylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'