missing translation for 'onlineSavingsMsg'
Learn More

4-Bromotoluene, 98%

Product Code. 11324769
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324769 25 g 25g
11472528 100 g 100g
11482528 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324769 Supplier Thermo Scientific Alfa Aesar Supplier No. A15843.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-38-7 | C7H7Br | 171.04 g/mol

4-Bromotoluene was used in the synthesis of ketones by Cu/Pd-catalyzed decarboxylative cross-coupling reaction. 4-Bromotoluene undergoes Heck reaction with styrene in the presence of in situ generated palladium complexes of phosphine-functionalized N-heterocyclic carbene ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromotoluene was used in the synthesis of ketones by Cu/Pd-catalyzed decarboxylative cross-coupling reaction. 4-Bromotoluene undergoes Heck reaction with styrene in the presence of in situ generated palladium complexes of phosphine-functionalized N-heterocyclic carbene ligands.

Solubility
Insoluble in water. Soluble in methanol.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-38-7
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.04
MDL Number MFCD00000109
InChI Key ZBTMRBYMKUEVEU-UHFFFAOYSA-N
Synonym 4-bromotoluene, p-bromotoluene, 4-methylbromobenzene, p-tolyl bromide, benzene, 1-bromo-4-methyl, toluene, p-bromo, p-methylphenyl bromide, p-methylbromobenzene, 1-methyl-4-bromobenzene, 4-bromo-1-methylbenzene
PubChem CID 7805
IUPAC Name 1-bromo-4-methylbenzene
SMILES CC1=CC=C(Br)C=C1

Specifications

Melting Point 26°C to 29°C
Density 1.41
Boiling Point 183°C to 184°C
Flash Point 85°C (185°F)
Quantity 25 g
Beilstein 1903636
Merck Index 14,1439
Solubility Information Insoluble in water. Soluble in methanol.
Formula Weight 171.04
Percent Purity 98%
Chemical Name or Material 4-Bromotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.