missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothiophenol, 98%

Product Code. 11438843
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438843 50 g 50g
11428843 10 g 10g
11448843 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438843 Supplier Thermo Scientific Alfa Aesar Supplier No. A10555.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-53-6 | C6H5BrS | 189.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-53-6
Molecular Formula C6H5BrS
Molecular Weight (g/mol) 189.07
MDL Number MFCD00004845
InChI Key FTBCOQFMQSTCQQ-UHFFFAOYSA-N
Synonym 4-bromothiophenol, benzenethiol, 4-bromo, p-bromothiophenol, p-bromobenzenethiol, 4-bromobenzene-1-thiol, 4-bromo-benzenethiol, 4-bromo thiophenol, p-bromophenyl thiol, benzenethiol, p-bromo, unii-p3wuo03x17
PubChem CID 66049
IUPAC Name 4-bromobenzenethiol
SMILES C1=CC(=CC=C1S)Br

Specifications

Melting Point 72°C to 78°C
Boiling Point 239°C
Odor Unpleasant
Quantity 50 g
UN Number UN2923
Beilstein 507031
Sensitivity Air Sensitive
Formula Weight 189.08
Percent Purity 98%
Chemical Name or Material 4-Bromothiophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.