missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothiophene-2-carboxaldehyde, 96%

Product Code. 11490893
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490893 100 g 100g
11470893 5 g 5g
11480893 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490893 Supplier Thermo Scientific Alfa Aesar Supplier No. A13304.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18791-75-8 | C5H3BrOS | 191.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18791-75-8
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00005431
InChI Key PDONIKHDXYHTLS-UHFFFAOYSA-N
Synonym 4-bromothiophene-2-carboxaldehyde, 4-bromo-2-thiophenecarboxaldehyde, 4-bromo-2-formylthiophene, 4-bromo-2-thiophenecarbaldehyde, 4-bromo-thiophene-2-carbaldehyde, 4-bromo-2-thiophene carboxaldehyde, 4-bromothiophen-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 4-bromo, 4-bromothiophene-2-aldehyde, 3-bromothiophene-5-carboxaldehyde
PubChem CID 87792
IUPAC Name 4-bromothiophene-2-carbaldehyde
SMILES BrC1=CSC(C=O)=C1

Specifications

Melting Point 44°C to 48°C
Boiling Point 114°C to 115°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Sensitivity Air sensitive
Formula Weight 191.05
Percent Purity 96%
Chemical Name or Material 4-Bromothiophene-2-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.