missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothiobenzamide, 97%, Thermo Scientific Chemicals

Catalog Number 15462358
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
26197-93-3
C7H6BrNS
216.096
MFCD04114360
AIPANIYQEBQYGC-UHFFFAOYSA-N
4-bromothiobenzamide, 4-bromo-thiobenzamide, p-bromothiobenzamide, 4-bromobenzothioamide, benzenecarbothioamide, 4-bromo, 4-bromobenzene-1-carbothioamide, amino 4-bromophenyl methane-1-thione, acmc-1cehr, 4-bromanylbenzenecarbothioamide
1549289
4-bromobenzenecarbothioamide
C1=CC(=CC=C1C(=S)N)Br
This item is not returnable. View return policy
26197-93-3
C7H6BrNS
216.096
MFCD04114360
AIPANIYQEBQYGC-UHFFFAOYSA-N
4-bromothiobenzamide, 4-bromo-thiobenzamide, p-bromothiobenzamide, 4-bromobenzothioamide, benzenecarbothioamide, 4-bromo, 4-bromobenzene-1-carbothioamide, amino 4-bromophenyl methane-1-thione, acmc-1cehr, 4-bromanylbenzenecarbothioamide
1549289
4-bromobenzenecarbothioamide
C1=CC(=CC=C1C(=S)N)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 139°C to 143°C
Color Brown to White
Quantity 1 g
Formula Weight 216.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Bromothiobenzamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'