missing translation for 'onlineSavingsMsg'
Learn More

4-Bromostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

Product Code. 11459006
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11459006

Brand: Thermo Scientific Alfa Aesar A17896.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2039-82-9 | C8H7Br | 183.05 g/mol

4-Bromostyrene is widely utilized in the synthesis of nitroolefins by alkene cross-metathesis. It plays an important role in the Heck reaction to the synthesis of poly(1,4-phenylenevinylene). It is employed in the structure activity relationships (SAR) study of the chemical and biochemical properties of the vinyl group of styrene. It is also used to investigate the photochemical growth of Br-terminated self-assembled monolayers. It is also involved in the synthesis of silsesquioxanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromostyrene is widely utilized in the synthesis of nitroolefins by alkene cross-metathesis. It plays an important role in the Heck reaction to the synthesis of poly(1,4-phenylenevinylene). It is employed in the structure activity relationships (SAR) study of the chemical and biochemical properties of the vinyl group of styrene. It is also used to investigate the photochemical growth of Br-terminated self-assembled monolayers. It is also involved in the synthesis of silsesquioxanes.

Solubility
Miscible with ethanol, ether and benzene.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, bases, oxidizing agents and halogens. Keep away from sources of ignition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2039-82-9
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.05
MDL Number MFCD00000110
InChI Key WGGLDBIZIQMEGH-UHFFFAOYSA-N
Synonym 4-bromostyrene, 1-bromo-4-vinylbenzene, p-bromostyrene, benzene, 1-bromo-4-ethenyl, styrene, p-bromo, 1-4-bromophenyl ethylene, unii-tvk4s80xlu, p-bromo styrene, 1-bromo-4-vinyl-benzene, tvk4s80xlu
PubChem CID 16263
IUPAC Name 1-bromo-4-ethenylbenzene
SMILES BrC1=CC=C(C=C)C=C1

Specifications

Melting Point 3°C to 6°C
Density 1.4
Boiling Point 88°C to 89°C (16 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.594
Quantity 25 g
Beilstein 1634204
Solubility Information Miscible with ethanol,ether and benzene.
Formula Weight 183.05
Percent Purity 98%
Chemical Name or Material 4-Bromostyrene, stabilized with 0.1% 4-tert-butylcatechol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.