missing translation for 'onlineSavingsMsg'
Learn More

4-Bromosalicylaldehyde, 97%

Product Code. 15481018
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15481018 5 g 5g
15491018 250 mg 250mg
15471018 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481018 Supplier Thermo Scientific Alfa Aesar Supplier No. H54850.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22532-62-3 | C7H5BrO2 | 201.019 g/mol

It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in methanol.

Notes
Strong bases and oxidizing agents are to be avoided.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22532-62-3
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD06252606
InChI Key HXTWKHXDFATMSP-UHFFFAOYSA-N
Synonym 4-bromosalicylaldehyde, 4-bromo-2-hydroxy-benzaldehyde, 2-hydroxy-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-hydroxy, 4-bromo-2-hydroxybenzaldheyde, 4-bromosalicyladehyde, pubchem16944, acmc-1cq5t, intermediates-zcf02605, 4-bromo-2-hydroxybenaldehyde
PubChem CID 4066019
IUPAC Name 4-bromo-2-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1Br)O)C=O

Specifications

Melting Point 50°C to 54°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3077
Solubility Information Soluble in methanol.
Formula Weight 201.02
Percent Purity 97%
Chemical Name or Material 4-Bromosalicylaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.