missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoresorcinol, 98%

Product Code. p-3777698 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10112900

Brand: Thermo Scientific Acros 107430050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6626-15-9 | C6H5BrO2 | 189.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6626-15-9
Molecular Formula C6H5BrO2
Molecular Weight (g/mol) 189.01
MDL Number MFCD00002272
InChI Key MPCCNXGZCOXPMG-UHFFFAOYSA-N
Synonym 4-bromoresorcinol, 1-bromo-2,4-dihydroxybenzene, 1,3-benzenediol, 4-bromo, 4-bromo-1,3-dihydroxybenzene, 4-bromo-benzene-1,3-diol, 4-bromo-resorcinol, resorcinol, 4-bromo, acmc-209nuf, 2,4-dihydroxybromobenzene
PubChem CID 81105
IUPAC Name 4-bromobenzene-1,3-diol
SMILES C1=CC(=C(C=C1O)O)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 99°C to 102°C
Color Beige to Pink
Boiling Point 150°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(OH)2
Quantity 5 g
Beilstein 06, 821
Formula Weight 189.01
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 4-Bromoresorcinol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.