missing translation for 'onlineSavingsMsg'
Learn More

4-Bromopyrazole, 98%

Product Code. 10386344 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10386344 5 g Glass bottle 5g
10673975 1 g Glass bottle 1g
10245054 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10386344 Supplier Thermo Scientific Acros Supplier No. 396820050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2075-45-8 | C3H3BrN2 | 146.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2075-45-8
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.98
MDL Number MFCD00075602
InChI Key WVGCPEDBFHEHEZ-UHFFFAOYSA-N
PubChem CID 16375
IUPAC Name 4-bromo-1H-pyrazole
SMILES BrC1=CNN=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 93.0°C to 96.0°C
Color White to Cream
Boiling Point 250.0°C to 260.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Synonym 4-bromopyrazole, 1h-pyrazole, 4-bromo, 4-brompyrazole, 4-bromo-pyrazole, pyrazole, 4-bromo, 4-bromo pyrazole, byz, zlchem 126, 1h-pyrazole,4-bromo, pubchem11041
Formula Weight 146.97
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromopyrazole
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromopyrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.