missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromopropiophenone, TRC

Product Code. 30335164
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
missing translation for 'unitSize'
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335164 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30335164 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB68724510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4'-Bromopropiophenone
CAS 10342-83-3
Molecular Formula C9H9BrO
Quantity 10 g
Synonym 1-(4-Bromophenyl)-1-propanone, 4'-Bromopropiophenone, 1-(4-Bromophenyl)propanone, 4-Bromophenyl Ethyl Ketone, 4-Ethylcarbonyl-1-bromobenzene, Ethyl 4-Bromophenyl Ketone, NSC 83574, p-Bromopropiophenone
InChI Formula InChI=1S/C9H9BrO/c1-2-9(11)7-3-5-8(10)6-4-7/h3-6H,2H2,1H3
SMILES CCC(=O)c1ccc(Br)cc1
IUPAC Name 1-(4-bromophenyl)propan-1-one
Molecular Weight (g/mol) 213.07
Formula Weight 211.9837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.