missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromopropiophenone, 97%

Product Code. 11334769
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Unit Size:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334769 25 g 25g
11406917 50 g 50g
11416917 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334769 Supplier Thermo Scientific Alfa Aesar Supplier No. A14349.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10342-83-3 | C9H9BrO | 213.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10342-83-3
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.07
MDL Number MFCD00000106
InChI Key UOMOSYFPKGQIKI-UHFFFAOYSA-N
Synonym 4'-bromopropiophenone, 1-4-bromophenyl propan-1-one, p-bromopropiophenone, 4-bromopropiophenone, propiophenone, 4'-bromo, 4-bromophenyl ethyl ketone, 1-propanone, 1-4-bromophenyl, 1-4-bromophenyl-1-propanone, aurora 22532
PubChem CID 66312
IUPAC Name 1-(4-bromophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 45°C to 47°C
Boiling Point 129°C to 131°C (7 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1210311
Formula Weight 213.08
Percent Purity 97%
Chemical Name or Material 4'-Bromopropiophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.