missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromophthalic acid, 98%, Thermo Scientific™
£25.48 - £88.40
Chemical Identifiers
CAS | 6968-28-1 |
---|---|
Molecular Formula | C8H5BrO4 |
Molecular Weight (g/mol) | 245.028 |
MDL Number | MFCD00043538 |
InChI Key | AZXKGUVDIORSED-UHFFFAOYSA-N |
Synonym | 4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid |
PubChem CID | 81428 |
IUPAC Name | 4-bromophthalic acid |
SMILES | C1=CC(=C(C=C1Br)C(=O)O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11313047
|
Thermo Scientific Alfa Aesar
L13418.03 |
1g |
£25.48
1g |
Estimated Shipment: 07-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11323047
|
Thermo Scientific Alfa Aesar
L13418.06 |
5g |
£88.40
5g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
6968-28-1 | |
245.028 | |
AZXKGUVDIORSED-UHFFFAOYSA-N | |
81428 | |
C1=CC(=C(C=C1Br)C(=O)O)C(=O)O |
C8H5BrO4 | |
MFCD00043538 | |
4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid | |
4-bromophthalic acid |
Specifications
4-Bromophthalic acid | |
162°C to 164°C | |
MFCD00043538 | |
4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid | |
AZXKGUVDIORSED-UHFFFAOYSA-N | |
4-bromophthalic acid | |
81428 | |
98% |
6968-28-1 | |
C8H5BrO4 | |
1959086 | |
1g | |
C1=CC(=C(C=C1Br)C(=O)O)C(=O)O | |
245.028 | |
245.03 |
RUO – Research Use Only