missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophthalic acid, 98%, Thermo Scientific Chemicals

Product Code. 11323047
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11323047

Brand: Thermo Scientific Alfa Aesar L13418.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6968-28-1
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00043538
InChI Key AZXKGUVDIORSED-UHFFFAOYSA-N
Synonym 4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid
PubChem CID 81428
IUPAC Name 4-bromophthalic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)C(=O)O

Specifications

Melting Point 162°C to 164°C
Quantity 5 g
Beilstein 1959086
Formula Weight 245.03
Percent Purity 98%
Chemical Name or Material 4-Bromophthalic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.