missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetic acid, 99%

Product Code. 11428226
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428226 100 g 100g
11408226 5 g 5g
11418226 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428226 Supplier Thermo Scientific Alfa Aesar Supplier No. A15325.22
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1878-68-8 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-68-8
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00004342
InChI Key QOWSWEBLNVACCL-UHFFFAOYSA-N
Synonym 4-bromophenylacetic acid, 2-4-bromophenyl acetic acid, 4-bromophenyl acetic acid, p-bromophenylacetic acid, 4-bromobenzeneacetic acid, benzeneacetic acid, 4-bromo, p-bromophenyl acetic acid, acetic acid, p-bromophenyl, p-bromphenylessigsaure german
PubChem CID 74654
ChEBI CHEBI:1790
IUPAC Name 2-(4-bromophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(Br)C=C1

Specifications

Melting Point 115°C to 119°C
Quantity 100 g
Beilstein 972146
Formula Weight 215.05
Percent Purity 99%
Chemical Name or Material 4-Bromophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.