missing translation for 'onlineSavingsMsg'
Learn More

4-bromophenyl isothiocyanate, Thermo Scientific™

Product Code. 10779025
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10779025 25 g 25g
10211003 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10779025 Supplier Thermo Scientific Maybridge Supplier No. SB01925EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1985-12-2 , 125-20-2
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
InChI Key XQACWEBGSZBLRG-UHFFFAOYSA-N
Synonym 4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate
PubChem CID 16133
IUPAC Name 1-bromo-4-isothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)Br

Specifications

Assay Percent Range 97%
Quantity 25 g
Formula Weight 214.09
Percent Purity 97%
Chemical Name or Material 4-Bromophenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.