missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromophenyl)cyclopropylmethanone, 95%

Product Code. 10279293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10279293 250 g 250g
10730414 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10279293 Supplier Thermo Scientific Acros Supplier No. 235422500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6952-89-2 | C10H9BrO | 225.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6952-89-2
MDL Number MFCD00019227
InChI Key QTHHOINSCNBYQO-UHFFFAOYSA-N
Synonym 4-bromophenyl cyclopropyl methanone, 4-bromophenyl cyclopropyl ketone, 4-bromophenyl cyclopropylmethanone, methanone, 4-bromophenyl cyclopropyl, 4-bromo-cyclopropylcarbonylbenzene, acmc-1bag8, 4-bromobenzoyl cyclopropane, bromophenylcyclopropylmethanone
PubChem CID 81394
IUPAC Name (4-bromophenyl)-cyclopropylmethanone
SMILES C1CC1C(=O)C2=CC=C(C=C2)Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Beige to Orange or Yellow
Density 1.4500g/mL
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C10H9BrO
Refractive Index 1.5919
Quantity 250 g
Specific Gravity 1.45
Molecular Weight (g/mol) 225.09
Formula Weight 225.09
Percent Purity 95%
Physical Form Crystalline Powder or Crystalline Low Melting Mass
Chemical Name or Material (4-Bromophenyl)cyclopropylmethanone
Show More Show Less
compliance-icons
Product Identifier
  • (4-Bromophenyl)cyclopropylmethanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.