missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl chloroformate, 95+%

Product Code. p-4228468 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10779078 1 g Glass bottle 1g
10126357 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10779078 Supplier Thermo Scientific Acros Supplier No. 430150010
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7693-44-9 | C7H4BrClO2 | 235.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7693-44-9
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.46
MDL Number MFCD00013256
InChI Key IKMNJYGTSSQNSE-UHFFFAOYSA-N
Synonym 4-bromophenyl chloroformate, acmc-1bh45, 4-bromophenylcarbonochloridate, 4-bromophenyl carbonochloridate, carbonochloridic acid, 4-bromophenyl ester, 4-bromophenyl carbonochloridate;4-bromophenyl carbonochloridate
PubChem CID 5237736
IUPAC Name (4-bromophenyl) carbonochloridate
SMILES C1=CC(=CC=C1OC(=O)Cl)Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Density 1.6480g/mL
Boiling Point 118°C (16.0 hPa)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.648
Formula Weight 235.46
Percent Purity 95+%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Bromophenyl chloroformate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Bromophenyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.