missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl acetate, 99%, Thermo Scientific™

Product Code. 15259836 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15259836 5 g 5g
15269836 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15259836 Supplier Thermo Scientific Acros Supplier No. 452770050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1927-95-3
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
InChI Key XEXHCQJRVMVJMY-UHFFFAOYSA-N
Synonym 4-bromophenol acetate, p-bromophenyl acetate, phenol, 4-bromo-, acetate, p-acetoxybromobenzene, 4-bromoacetoxybenzene, phenol, 4-bromo-, 1-acetate, 4-bromophenyl acetate, phenol, p-bromo-, acetate, phenol, 4-bromo-,1-acetate, acetic acid 4-bromophenyl ester
PubChem CID 74726
IUPAC Name (4-bromophenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)Br

Specifications

Assay Percent Range 98.5% min. (GC)
Quantity 5 g
Formula Weight 215.05
Percent Purity 99%
Chemical Name or Material 4-Bromophenyl acetate
compliance-icons compliance-icons
Product Identifier
  • 4-Bromophenyl acetate, 99%
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.