missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromophenyl acetate, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Assay Percent Range | 98.5% min. (GC) |
Quantity | 25 g |
Formula Weight | 215.05 |
Percent Purity | 99% |
Chemical Name or Material | 4-Bromophenyl acetate |
Safety and Handling
Product Identifier
- 4-Bromophenyl acetate, 99%
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H317-May cause an allergic skin reaction.
- H411-Toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'