missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (4-Bromophenoxy)-tert-butyldimethylsilane, 97%

Product Code. 15462298
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462298 5 mL 5mL
15472298 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462298 Supplier Alfa Aesar™ Supplier No. H55902.AA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 67963-68-2
Molecular Formula C12H19BrOSi
Molecular Weight (g/mol) 287.272
MDL Number MFCD00239382
InChI Key DLGZGLKSNRKLSM-UHFFFAOYSA-N
Synonym 4-bromophenoxy tert-butyl dimethylsilane, 4-bromophenoxy-tert-butyldimethylsilane, p-bromophenoxy t-butyl dimethylsilane, 4-bromophenoxy-tert-butyl-dimethylsilane, 1-bromo-4-tert-butyldimethylsiloxy benzene, 4-bromophenol tert-butyldimethylsilyl ether, silane, 4-bromophenoxy 1,1-dimethylethyl dimethyl, 1-bromo-4-1,1-dimethylethyl dimethylsilyl oxy benzene, acmc-1b8oo
PubChem CID 4227167
IUPAC Name (4-bromophenoxy)-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OC1=CC=C(C=C1)Br

Specifications

Boiling Point 137°C (25mmHg)
Color Colorless
Physical Form Liquid
Quantity 5 mL
UN Number UN1993
Formula Weight 287.27
Percent Purity 97%
Refractive Index 1.5110
Density 1.174g/mL
Chemical Name or Material (4-Bromophenoxy)-tert-butyldimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.