missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenol, 99%, Thermo Scientific Chemicals

Catalog Number 11442667
Click to view available options
Quantity:
100 g
500 g
2500 g
106-41-2
C6H5BrO
173.009
MFCD00002313
GZFGOTFRPZRKDS-UHFFFAOYSA-N
p-bromophenol, phenol, 4-bromo, p-bromohydroxybenzene, 4-bromo-phenol, p-bromophenic acid, phenol, p-bromo, para-bromophenol, 4-bromo phenol, unii-lao4j0183i, ccris 632
7808
4-bromophenol
C1=CC(=CC=C1O)Br
106-41-2
C6H5BrO
173.009
MFCD00002313
GZFGOTFRPZRKDS-UHFFFAOYSA-N
p-bromophenol, phenol, 4-bromo, p-bromohydroxybenzene, 4-bromo-phenol, p-bromophenic acid, phenol, p-bromo, para-bromophenol, 4-bromo phenol, unii-lao4j0183i, ccris 632
7808
4-bromophenol
C1=CC(=CC=C1O)Br

4-Bromophenol is used as an intermediate in medicine grade. It is also used as a phenol that is used in Stille reactions. It is used in the intermediate of organic synthesis, flame retardant and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenol is used as an intermediate in medicine grade. It is also used as a phenol that is used in Stille reactions. It is used in the intermediate of organic synthesis, flame retardant and pharmaceutical industry.

Solubility
It is insoluble in water. It is soluble in 5%ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Store under inert gas. Air sensitive. Keep away from Oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 62°C to 66°C
Density 1.840
Boiling Point 235°C to 238°C
Flash Point >110°C (230°F)
Assay Percent Range 99%
Quantity 100 g
UN Number UN2811
Beilstein 1680024
Sensitivity Light sensitive
Merck Index 14,1428
Solubility Information It is insoluble in water. It is soluble in 5%ethanol.
Formula Weight 173.01
Percent Purity 99%
Chemical Name or Material 4-Bromophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'