missing translation for 'onlineSavingsMsg'
Learn More

4-Bromonaphthalene-1-boronic acid, 97%, Thermo Scientific™

Catalog Number 15471938
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
145965-14-6
C10H8BBrO2
250.89
MFCD08701764
DKKOMUVRCPKTFX-UHFFFAOYSA-N
4-bromonaphthalen-1-yl boronic acid, 4-bromonaphthalene-1-boronic acid, 4-bromo-1-naphthaleneboronic acid, boronic acid, 4-bromo-1-naphthalenyl, 4-bromo-1-naphthalenyl-boronic acid, acmc-209cw4, ksc524o6f, 4-bromo-1-naphthylboronic acid, 1-bromonaphthalene-4-boronic acid, 1-bromo-4-dihydroxyboryl naphthalene
23005424
(4-bromonaphthalen-1-yl)boronic acid
OB(O)C1=C2C=CC=CC2=C(Br)C=C1
This item is not returnable. View return policy
145965-14-6
C10H8BBrO2
250.89
MFCD08701764
DKKOMUVRCPKTFX-UHFFFAOYSA-N
4-bromonaphthalen-1-yl boronic acid, 4-bromonaphthalene-1-boronic acid, 4-bromo-1-naphthaleneboronic acid, boronic acid, 4-bromo-1-naphthalenyl, 4-bromo-1-naphthalenyl-boronic acid, acmc-209cw4, ksc524o6f, 4-bromo-1-naphthylboronic acid, 1-bromonaphthalene-4-boronic acid, 1-bromo-4-dihydroxyboryl naphthalene
23005424
(4-bromonaphthalen-1-yl)boronic acid
OB(O)C1=C2C=CC=CC2=C(Br)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 230°C to 260°C
Color Brown to White
Quantity 250 mg
Formula Weight 250.88
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Bromonaphthalene-1-boronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'