missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)tetrahydropyran, 97%, Thermo Scientific™

Product Code. 10356564
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10356564 10 g Amber Glass Bottle 10g
10378022 1 g Amber Glass Bottle 1g
10622855 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10356564 Supplier Thermo Scientific Maybridge Supplier No. CC29908EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 125552-89-8
Molecular Formula C6H11BrO
Molecular Weight (g/mol) 179.057
InChI Key LMOOYAKLEOGKJR-UHFFFAOYSA-N
Synonym 4-bromomethyl tetrahydropyran, 4-bromomethyltetrahydropyran, 4-bromomethyl oxane, 4-bromomethyl tetrahydro-2h-pyran, 4-bromomethyl-tetrahydro-pyran, 4-bromomethyl tetrahydro-pyran, 4-bromomethyl-tetrahydro-2h-pyran, 4-bromomethyl-tetrahydropyran, 2h-pyran, 4-bromomethyl tetrahydro
PubChem CID 2773286
IUPAC Name 4-(bromomethyl)oxane
SMILES C1COCCC1CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 179.06
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)tetrahydropyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.