missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)phenylacetic acid, 97%

Product Code. 11372889
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372889 1 g 1g
11382889 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372889 Supplier Thermo Scientific Alfa Aesar Supplier No. H27744.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 13737-36-5 | C9H9BrO2 | 229.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13737-36-5
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00010632
InChI Key WCOCCXZFEJGHTC-UHFFFAOYSA-N
Synonym 4-bromomethyl phenylacetic acid, 4-bromomethylphenylacetic acid, 2-4-bromomethyl phenyl acetic acid, pam acid, br-pam-linker, 4-carboxymethyl benzyl bromide, 4-bromomethyl phenyl acetic acid, 4-bromomethyl phenylaceticacid
PubChem CID 4519056
IUPAC Name 2-[4-(bromomethyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)CBr

Specifications

Melting Point 178°C to 180°C
Quantity 1 g
UN Number UN3261
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)phenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.