missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)biphenyl, 96%

Product Code. 10724953 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10724953 5 g 5g
10120912 25 g 25g
11924531 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10724953 Supplier Thermo Scientific Acros Supplier No. 368950050
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2567-29-5 | C13H11Br | 247.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2567-29-5
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.13
InChI Key HZQLUIZFUXNFHK-UHFFFAOYSA-N
Synonym 4-bromomethylbiphenyl, 4-bromomethyl biphenyl, 4-bromomethyl-1,1'-biphenyl, 4-phenylbenzyl bromide, 1-bromomethyl-4-phenylbenzene, bromoethylbiphenyl, 4-biphenylmethylbromide, 4-bromomethyl-biphenyl
PubChem CID 257716
IUPAC Name 1-(bromomethyl)-4-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CBr

Specifications

Melting Point 83.0°C to 86.0°C
Color Beige to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H4CH2Br
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene
Formula Weight 247.13
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-(Bromomethyl)biphenyl
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Bromomethyl)biphenyl
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.