missing translation for 'onlineSavingsMsg'
Learn More

4’-(Bromomethyl)biphenyl-2-carboxylic Acid tert-Butyl Ester, TRC

Product Code. 30331349
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331349 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331349 Supplier Toronto Research Chemicals Supplier No. TRCB6853851G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Dimethylethyl 4'-(Bromomethyl)biphenyl-2-carboxylate
CAS 114772-40-6
Molecular Formula C18 H19 Br O2
Quantity 1 g
Synonym 1,1-Dimethylethyl 4'-(Bromomethyl)-biphenyl-2-carboxylate, 1,1-Dimethylethyl 4'-(bromomethyl)biphenyl-2-carboxylate, 2-(4-Bromomethylphenyl)benzoic acid tert-butyl ester, 2'-(tert-Butoxylcarbonyl)biphenyl-4-ylmethyl bromide, 4-(Bromomethyl)-2'-(tert-butoxycarbonyl)biphenyl, 4'-(Bromomethyl)-2-(tert-butoxycarbonyl)biphenyl, 4'-(Bromomethyl)[1,1'-biphenyl]-2-carboxylic acid 1,1-dimethylethyl ester, 4'-(Bromomethyl)biphenyl-2-carboxylic acid tert-butyl ester, 4'-Bromomethyl[1,1'-biphenyl]-2-carboxylic acid tert-butyl ester, 4'-Bromomethylbiphenyl-2-carboxylic Acid tert-Butyl Ester, tert-Butyl 2-(4-bromomethylphenyl)benzoate, tert-Butyl 4-(bromomethyl)biphenyl-2'-carboxylate, tert-Butyl 4-bromomethylbiphenyl-2-carboxylate, tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate, Telmisartan Imp H (EP)
InChI Formula InChI=1S/C18H19BrO2/c1-18(2,3)21-17(20)16-7-5-4-6-15(16)14-10-8-13(12-19)9-11-14/h4-11H,12H2,1-3H3
SMILES CC(C)(C)OC(=O)c1ccccc1c2ccc(CBr)cc2
IUPAC Name tert-butyl 2-[4-(bromomethyl)phenyl]benzoate
Molecular Weight (g/mol) 347.25
Formula Weight 346.0568
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.