missing translation for 'onlineSavingsMsg'
Learn More

[4-(Bromomethyl)benzyl]triphenylphosphonium bromide, 98%, Thermo Scientific™

Product Code. p-7023725
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498187 25 g 25g
11408197 100 g 100g
11418197 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498187 Supplier Thermo Scientific Alfa Aesar Supplier No. A17563.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14366-74-6
Molecular Formula C26H23Br2P
Molecular Weight (g/mol) 526.25
MDL Number MFCD00031635
InChI Key KUXBEOGHKWNPTG-UHFFFAOYSA-M
Synonym 4-bromomethyl benzyl triphenylphosphonium bromide, p-bromomethyl benzyltriphenylphosphonium bromide, phosphonium, p-bromomethylbenzyl triphenyl-, bromide, acmc-1c314, wln: e1r d1pr&r&r &e, 4-bromomethyl benzyl triphenylphosphoniumbromide, 4-bromomethyl phenyl methyl-triphenylphosphanium bromide, 4-bromomethyl phenyl methyl triphenylphosphanium bromide, phosphonium, 4-bromomethyl phenyl methyl triphenyl-,bromide 1:1
PubChem CID 197927
IUPAC Name [4-(bromomethyl)phenyl]methyl-triphenylphosphanium;bromide
SMILES [Br-].BrCC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 246°C to 248°C (decomposition)
Quantity 25 g
Beilstein 3820074
Formula Weight 526.25
Percent Purity 98%
Chemical Name or Material [4-(Bromomethyl)benzyl]triphenylphosphonium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.