missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)benzonitrile, 98%

Product Code. 11499077
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499077 5 g 5g
11409087 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499077 Supplier Thermo Scientific Alfa Aesar Supplier No. B20641.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17201-43-3 | C8H6BrN | 196.047 g/mol

4-(Bromomethyl)benzonitrile is used in the synthesis of ligands containing a chelating pyrazolyl-pyridine group with a pendant aromatic nitrile. It is also useful in the preparation of 4-[(2H-tetra-zol-2-yl)methyl]benzonitrile by reaction with 2H-tetrazole in the presence of potassium hydroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Bromomethyl)benzonitrile is used in the synthesis of ligands containing a chelating pyrazolyl-pyridine group with a pendant aromatic nitrile. It is also useful in the preparation of 4-[(2H-tetra-zol-2-yl)methyl]benzonitrile by reaction with 2H-tetrazole in the presence of potassium hydroxide.

Solubility
Soluble in chloroform and methanol. Insoluble in water.

Notes
Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17201-43-3
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00001829
InChI Key UMLFTCYAQPPZER-UHFFFAOYSA-N
Synonym 4-cyanobenzyl bromide, 4-bromomethyl benzonitrile, p-cyanobenzyl bromide, alpha-bromo-p-tolunitrile, benzonitrile, 4-bromomethyl, 4-bromomethyl-benzonitrile, alpha-bromo-p-toluonitrile, p-bromomethyl benzonitrile, 4-cyano benzyl bromide, benzonitrile, p-bromomethyl
PubChem CID 86996
IUPAC Name 4-(bromomethyl)benzonitrile
SMILES C1=CC(=CC=C1CBr)C#N

Specifications

Melting Point 113°C to 117°C
Quantity 5 g
UN Number UN2923
Beilstein 636717
Solubility Information Soluble in chloroform and methanol. Insoluble in water.
Formula Weight 196.05
Percent Purity 98%
Chemical Name or Material 4-(Bromomethyl)benzonitrile
compliance-icons
Product Identifier
  • 4-(Bromomethyl)benzonitrile
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 4-(Bromomethyl)benzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.