missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Bromomethyl)benzoic Acid Methyl Ester, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 4-(Bromomethyl)benzoic Acid Methyl Ester |
| CAS | 2417-72-3 |
| Molecular Formula | C9H9BrO2 |
| Quantity | 10 g |
| Synonym | 4-(Carbomethoxy)benzyl Bromide, 4-(Methoxycarbonyl)benzyl Bromide, Methyl 4-(bromomethyl)benzoate, Methyl p-(Bromomethyl)benzoate, Methyl alpha-bromo-p-toluate, NSC 83959, p-(Bromomethyl)benzoic Acid Methyl Ester, p-(Carbomethoxy)benzyl Bromide, p-(Methoxycarbonyl)benzyl Bromide, alpha-Bromo-p-toluic Acid Methyl Ester |
| InChI Formula | InChI=1S/C9H9BrO2/c1-12-9(11)8-4-2-7(6-10)3-5-8/h2-5H,6H2,1H3 |
| SMILES | COC(=O)c1ccc(CBr)cc1 |
| IUPAC Name | methyl 4-(bromomethyl)benzoate |
| Molecular Weight (g/mol) | 229.07 |
| Formula Weight | 227.9786 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction