missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)benzeneboronic acid, tech. 85%

Product Code. 11324326
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324326 5 g 5g
11314326 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324326 Supplier Thermo Scientific Alfa Aesar Supplier No. L19953.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68162-47-0 | C7H8BBrO2 | 214.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68162-47-0
Molecular Formula C7H8BBrO2
Molecular Weight (g/mol) 214.85
MDL Number MFCD01318113
InChI Key PDNOURKEZJZJNZ-UHFFFAOYSA-N
Synonym 4-bromomethyl phenylboronic acid, 4-bromomethylphenylboronic acid, 4-bromomethyl phenyl boronic acid, 4-bromomethyl benzeneboronic acid, 4-boronobenzyl bromide, p-bromomethyl phenylboronic acid, 4-bromomethylphenyl boronic acid, 4-bromomethylbenzeneboronic acid, boronic acid, 4-bromomethyl phenyl
PubChem CID 2735593
IUPAC Name [4-(bromomethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(CBr)C=C1

Specifications

Melting Point ∼145°C to 150°C
Quantity 5 g
UN Number UN3261
Beilstein 3030979
Formula Weight 214.85
Percent Purity 85%
Grade Technical
Chemical Name or Material 4-(Bromomethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.