missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)benzeneboronic acid pinacol ester, 95%

Product Code. 11334326
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334326 1 g 1g
11344326 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334326 Supplier Thermo Scientific Alfa Aesar Supplier No. L19954.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 138500-85-3 | C13H18BBrO2 | 297.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138500-85-3
Molecular Formula C13H18BBrO2
Molecular Weight (g/mol) 297.00
MDL Number MFCD02179493
InChI Key CBUOGMOTDGNEAW-UHFFFAOYSA-N
Synonym 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl benzeneboronic acid pinacol ester, 4-bromomethylphenylboronic acid pinacol ester, 4-bromomethylphenylboronic acid, pinacol ester, 4-bromomethyl phenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl, 2-4-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaboronate, 4-bromomethyl benzeneboronicacidpinacolester, 4-bromomethyl benzeneboronic acid, pinacol ester
PubChem CID 3734506
IUPAC Name 2-[4-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CBr)C=C1

Specifications

Melting Point 83°C to 85°C
Quantity 1 g
UN Number UN3261
Beilstein 7921808
Formula Weight 297
Percent Purity 95%
Chemical Name or Material 4-(Bromomethyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.