missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Bromomethyl)benzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Synonym | 4-bromomethyl benzaldehyde, 4-formylbenzyl bromide, alpha-bromo-4-formyltoluene, benzaldehyde,4-bromomethyl, benzaldehyde, 4-bromomethyl, pubchem19839, p-bromomethyl benzaldehyde, 4-bromomethy benzaldehyde, 4-bromomethyl-benzaldehyde |
Formula Weight | 199.05 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Bromomethyl)benzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'