missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethyl-2'-fluorobiphenyl, 97%

Product Code. 15496207
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15496207

Brand: Thermo Scientific Alfa Aesar H33396.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 193013-76-2 | C13H10BrF | 265.125 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193013-76-2
Molecular Formula C13H10BrF
Molecular Weight (g/mol) 265.125
MDL Number MFCD15146811
InChI Key SAUALJYDYRRWPW-UHFFFAOYSA-N
Synonym 4'-bromomethyl-2-fluoro-1,1'-biphenyl, 4-bromomethyl-2'-fluorobiphenyl, 2-fluoro-4'-bromomethylbiphenyl
PubChem CID 18680932
IUPAC Name 1-(bromomethyl)-4-(2-fluorophenyl)benzene
SMILES C1=CC=C(C(=C1)C2=CC=C(C=C2)CBr)F

Specifications

Melting Point 72°C to 74°C
Quantity 250 mg
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 265.13
Percent Purity 97%
Chemical Name or Material 4-Bromomethyl-2'-fluorobiphenyl

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.