missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromomethyl-2(1H)-quinolinone, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 4-Bromomethyl-2(1H)-quinolinone |
| CAS | 4876-10-2 |
| Molecular Formula | C10 H8 Br N O |
| Quantity | 5 g |
| Synonym | 4-(Bromomethyl)carbostyril, 4-(Bromomethyl)quinolin- 2(1H)-one, 4-Bromomethyl-1H-quinolin-2-one, NSC 108455 |
| InChI Formula | InChI=1S/C10H8BrNO/c11-6-7-5-10(13)12-9-4-2-1-3-8(7)9/h1-5H,6H2,(H,12,13) |
| SMILES | BrCC1=CC(=O)Nc2ccccc12 |
| IUPAC Name | 4-(bromomethyl)-1H-quinolin-2-one |
| Molecular Weight (g/mol) | 238.081 |
| Formula Weight | 236.979 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction