missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoisoxazole, 95%

Product Code. 15579573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15579573 1 g 1g
15589573 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15579573 Supplier Thermo Scientific Acros Supplier No. 458730010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97925-43-4 | C3H2BrNO | 147.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97925-43-4
Molecular Formula C3H2BrNO
Molecular Weight (g/mol) 147.96
InChI Key IRDVIGFQMLUJAM-UHFFFAOYSA-N
Synonym 4-bromoisoxazole, 4-bormoisoxazole, isoxazole, 4-bromo, isoxazole, 4-bromo-6ci,9ci, 4-bromo isoxazole, 4-bromo-isoxazole, zlchem 1329
PubChem CID 3862248
IUPAC Name 4-bromo-1,2-oxazole
SMILES C1=C(C=NO1)Br

Specifications

Melting Point 39.0°C to 41.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 147.96
Percent Purity 95%
Chemical Name or Material 4-Bromoisoxazole
compliance-icons
Product Identifier
  • 4-Bromoisoxazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.