missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoisoquinoline, 98%

Product Code. 11438506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438506 100 g 100g
11418506 5 g 5g
11428506 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438506 Supplier Thermo Scientific Alfa Aesar Supplier No. A15951.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1532-97-4 | C9H6BrN | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1532-97-4
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD00006904
InChI Key SCRBSGZBTHKAHU-UHFFFAOYSA-N
Synonym isoquinoline, 4-bromo, 4-bromo-isoquinoline, 4-isoquinolinyl bromide, 4-bromisochinolin, 4bromoisoquinoline, 4-bromoisoquinoline, 4-bromo isoquinoline, 4-bromoisoquinoline?, pubchem6212, acmc-1bu1h
PubChem CID 73743
IUPAC Name 4-bromoisoquinoline
SMILES BrC1=C2C=CC=CC2=CN=C1

Specifications

Melting Point 39°C to 43°C
Boiling Point 285°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 114431
Formula Weight 208.06
Percent Purity 98%
Chemical Name or Material 4-Bromoisoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.