missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoimidazole, 98%

Product Code. 15442409
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442409 5 g 5g
15452409 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442409 Supplier Thermo Scientific Alfa Aesar Supplier No. H64972.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2302-25-2 | C3H3BrN2 | 146.98 g/mol

Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2302-25-2
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.98
MDL Number MFCD00047021
InChI Key FHZALEJIENDROK-UHFFFAOYSA-N
Synonym 4-bromo-1h-imidazole, 4-bromoimidazole, 1h-imidazole, 4-bromo, 4 or 5-bromoimidazole, imidazole, 4-bromo, 4-bromo-3h-imidazole, imidazole, 4 or 5-bromo-6ci,7ci, 5-bromoimidazole, 3kqm, 4-bromo-imidazole
PubChem CID 96125
IUPAC Name 5-bromo-1H-imidazole
SMILES BrC1=CN=CN1

Specifications

Melting Point 131°C to 135°C
Quantity 5 g
UN Number UN2811
Beilstein 1615
Solubility Information Slightly soluble in water.
Formula Weight 146.97
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromoimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.