missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoheptane, 97%

Product Code. 11381717
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381717 5 g 5g
11391717 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381717 Supplier Thermo Scientific Alfa Aesar Supplier No. L04807.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 998-93-6 | C7H15Br | 179.10 g/mol

4-Bromoheptane is used in the preparation of 4-nitro-heptane by reacting with sodium nitrite as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoheptane is used in the preparation of 4-nitro-heptane by reacting with sodium nitrite as a reagent.

Solubility
Miscible with carbon tetrachloride, chloroform and benzene. Immiscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 998-93-6
Molecular Formula C7H15Br
Molecular Weight (g/mol) 179.10
MDL Number MFCD00039182
InChI Key BNUTXEKPXPZAIT-UHFFFAOYSA-N
Synonym heptane, 4-bromo, 4-bromo-heptane, acmc-20ammr, 4-bromoheptane 5g
PubChem CID 70460
IUPAC Name 4-bromoheptane
SMILES CCCC(Br)CCC

Specifications

Density 1.14
Boiling Point 60°C to 62°C (18 mmHg)
Flash Point 40°C (104°F)
Refractive Index 1.45
Quantity 5 g
UN Number UN1993
Beilstein 1697144
Solubility Information Miscible with carbon tetrachloride,chloroform and benzene. Immiscible with water.
Formula Weight 179.11
Percent Purity 97%
Chemical Name or Material 4-Bromoheptane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.