missing translation for 'onlineSavingsMsg'
Learn More

4-Bromofluorobenzene, 99%

Product Code. 10345880 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
10mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10345880 100 mL Glass bottle 100mL
10385830 10 mL Glass bottle 10mL
15730789 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10345880 Supplier Thermo Scientific Supplier No. 107011000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 460-00-4 | C6H4BrF | 175.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 460-00-4
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175.00
MDL Number MFCD00000342
InChI Key AITNMTXHTIIIBB-UHFFFAOYSA-N
Synonym 4-bromofluorobenzene, p-bromofluorobenzene, p-fluorobromobenzene, 4-fluorobromobenzene, benzene, 1-bromo-4-fluoro, 4-fluorophenyl bromide, 1-fluoro-4-bromobenzene, 4-fluoro-1-bromobenzene, p-fluorophenyl bromide, bromo fluorobenzene
PubChem CID 9993
IUPAC Name 1-bromo-4-fluorobenzene
SMILES FC1=CC=C(Br)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -8.0°C
Color Colorless
Density 1.5930g/mL
Boiling Point 150.0°C to 153.0°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H4F
Refractive Index 1.5260 to 1.5280
Quantity 100 mL
Beilstein 05, 209
Specific Gravity 1.593
Solubility Information Solubility in water: 0.14g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,acetone,soluble in ether
Formula Weight 175
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Bromofluorobenzene, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.