missing translation for 'onlineSavingsMsg'
Learn More

4-Bromodiphenyl ether, 99%

Product Code. 11361287
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361287 25 g 25g
11371287 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361287 Supplier Thermo Scientific Alfa Aesar Supplier No. L01473.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101-55-3 | C12H9BrO | 249.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-55-3
Molecular Formula C12H9BrO
Molecular Weight (g/mol) 249.107
MDL Number MFCD00000094
InChI Key JDUYPUMQALQRCN-UHFFFAOYSA-N
Synonym 4-bromodiphenyl ether, 4-bromophenyl phenyl ether, 4-bromophenoxybenzene, p-bromodiphenyl ether, p-phenoxybromobenzene, benzene, 1-bromo-4-phenoxy, p-bromophenyl phenyl ether, 4-bromodiphenylether, p-bromophenoxybenzene, ether, p-bromophenyl phenyl
PubChem CID 7565
ChEBI CHEBI:77421
IUPAC Name 1-bromo-4-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)Br

Specifications

Melting Point 17°C to 18°C
Density 1.42
Boiling Point 304°C to 305°C
Flash Point >110°C (230°F)
Refractive Index 1.608
Quantity 25 g
UN Number UN3082
Beilstein 1308972
Formula Weight 249.11
Percent Purity 99%
Chemical Name or Material 4-Bromodiphenyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.