missing translation for 'onlineSavingsMsg'
Learn More

4-Bromocrotonic acid benzoylmethyl ester, 97%, Thermo Scientific Chemicals

Product Code. 11393227
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393227 25 g 25g
11383227 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393227 Supplier Thermo Scientific Alfa Aesar Supplier No. L13765.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. Rückgaberichtlinie anzeigen

CAS: 154561-38-3 | Molecular Formula: C12H11BrO3 | Molecular Weight: 283.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 154561-38-3
Molecular Formula C12H11BrO3
Molecular Weight (g/mol) 283.12
MDL Number MFCD00236054
InChI Key JARTYZWEUJDLPS-UHFFFAOYSA-N
Synonym 2-oxo-2-phenylethyl 4-bromobut-2-enoate, acmc-20ajxj, 2-butenoic acid,4-bromo-, 2-oxo-2-phenylethyl ester
PubChem CID 53412107
IUPAC Name phenacyl 4-bromobut-2-enoate
SMILES BrCC=CC(=O)OCC(=O)C1=CC=CC=C1

Spezifikation

Melting Point 89°C to 91°C
Quantity 25 g
UN Number UN3335
Formula Weight 283.13
Percent Purity 97%
Chemical Name or Material 4-Bromocrotonic acid benzoylmethyl ester

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.