missing translation for 'onlineSavingsMsg'
Learn More

4-Bromocrotonic acid benzoylmethyl ester, 97%, Thermo Scientific Chemicals

Product Code. 11383227
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383227 5 g 5g
11393227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383227 Supplier Thermo Scientific Alfa Aesar Supplier No. L13765.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154561-38-3
Molecular Formula C12H11BrO3
Molecular Weight (g/mol) 283.12
MDL Number MFCD00236054
InChI Key JARTYZWEUJDLPS-UHFFFAOYSA-N
Synonym 2-oxo-2-phenylethyl 4-bromobut-2-enoate, acmc-20ajxj, 2-butenoic acid,4-bromo-, 2-oxo-2-phenylethyl ester
PubChem CID 53412107
IUPAC Name phenacyl 4-bromobut-2-enoate
SMILES BrCC=CC(=O)OCC(=O)C1=CC=CC=C1

Specifications

Melting Point 89°C to 91°C
Quantity 5 g
UN Number UN3335
Formula Weight 283.13
Percent Purity 97%
Chemical Name or Material 4-Bromocrotonic acid benzoylmethyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.