missing translation for 'onlineSavingsMsg'
Learn More

4-Bromocinnamic Acid, TRC

Product Code. 30343328
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30343328 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343328 Supplier Toronto Research Chemicals Supplier No. TRCB68243525G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromocinnamic Acid
CAS 1200-07-3
Molecular Formula C9H7BrO2
Quantity 25 g
InChI Formula InChI=1S/C9H7BrO2/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-6H,(H,11,12)/b6-3+
SMILES OC(=O)\C=C\c1ccc(Br)cc1
IUPAC Name (E)-3-(4-bromophenyl)prop-2-enoic acid
Molecular Weight (g/mol) 227.06
Formula Weight 225.9629
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.