missing translation for 'onlineSavingsMsg'
Learn More

4-Bromochlorobenzene, 99%

Product Code. 10102710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10102710 1 kg Plastic bottle 1kg
10627333 50 g Plastic bottle 50g
10148003 100 g Plastic bottle 100g
10519820 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10102710 Supplier Thermo Scientific Acros Supplier No. 106830010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-39-8 | C6H4BrCl | 191.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-39-8
Molecular Formula C6H4BrCl
Molecular Weight (g/mol) 191.45
MDL Number MFCD00000600
InChI Key NHDODQWIKUYWMW-UHFFFAOYSA-N
Synonym 4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene
PubChem CID 7806
IUPAC Name 1-bromo-4-chlorobenzene
SMILES ClC1=CC=C(Br)C=C1

Specifications

Melting Point 64.0°C to 67.0°C
CAS Min % 98.5
Color White
CAS Max % 100.0
Density 1.6510g/mL
Quantity 1 kg
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.651
Linear Formula BrC6H4Cl
Beilstein 05, 209
Boiling Point 196.0°C
Solubility Information Solubility in water: 0.045g/L (25°C). Other solubilities: soluble in most organic solvents
Formula Weight 191.45
Percent Purity 99%
Chemical Name or Material 4-Bromochlorobenzene, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.