Learn More
4-Bromobutyryl chloride, 97%
Description
4-Bromobutyryl chloride is used as a precursor for the synthesis of carolic acid by reacting with ethoxymagnesiomalonic ester. It is widely utilized as an intermediate in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Bromobutyryl chloride is used as a precursor for the synthesis of carolic acid by reacting with ethoxymagnesiomalonic ester. It is widely utilized as an intermediate in organic synthesis and pharmaceuticals.
Solubility
Miscible with organic solvents. Immiscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Remove all sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
| Density | 1.6 |
| Boiling Point | 102°C to 103°C (40 mmHg) |
| Flash Point | 91°C (195°F) |
| Linear Formula | Br(CH2)3COCl |
| Refractive Index | 1.492 |
| Quantity | 25 g |
| UN Number | UN3265 |
| Beilstein | 1743026 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Miscible with organic solvents. Immiscible with water. |
| Show More |
Safety and Handling
- 4-Bromobutyryl chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH014-Reacts violently with water.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: 4-Bromobutyryl chloride
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.