missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyryl chloride, 95%

Product Code. 10582841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10582841 5 g Glass bottle 5g
10351184 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10582841 Supplier Thermo Scientific Acros Supplier No. 303440050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 927-58-2 | C4H6BrClO | 185.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 927-58-2
Molecular Formula C4H6BrClO
Molecular Weight (g/mol) 185.46
MDL Number MFCD00000753
InChI Key LRTRXDSAJLSRTG-UHFFFAOYSA-N
Synonym 4-bromobutyryl chloride, butanoyl chloride, 4-bromo, g-bromobutyroyl chloride, butyryl chloride, 4-bromo, 4-bromobutyrylchloride, bromobutryl chloride, bromobutyryl chloride, acmc-20ammt, 4-bromobutanoylchloride, 4-bromobutyroylchloride
PubChem CID 70225
IUPAC Name 4-bromobutanoyl chloride
SMILES C(CC(=O)Cl)CBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow or Pink
Density 1.6020g/mL
Boiling Point 101°C (37.0 mmHg)
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)3COCl
Refractive Index 1.4910 to 1.494
Quantity 5 g
Specific Gravity 1.602
Formula Weight 185.46
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Bromobutyryl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.