missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyric acid, 97%

Product Code. 11415303
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415303 10 g 10g
11425303 50 g 50g
11435303 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11415303 Supplier Thermo Scientific Alfa Aesar Supplier No. A12870.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

CAS: 2623-87-2 | C4H7BrO2 | 167.00 g/mol

4-Bromobutyric acid is used as a bifunctional cross-linker and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobutyric acid is used as a bifunctional cross-linker and as a pharmaceutical intermediate.

Solubility
Soluble in water and dilute alkali.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature. It is sensitive to light. Incompatible with strong oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2623-87-2
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00002817
InChI Key GRHQDJDRGZFIPO-UHFFFAOYSA-N
Synonym 4-bromobutyric acid, butanoic acid, 4-bromo, 4-bromo-n-butyric acid, butyric acid, 4-bromo, gamma-bromobutyric acid, rarechem al bo 0176, 4-bromobutyricacid, carboxypropyl bromide, 4-bromo-butyric acid, pubchem3754
PubChem CID 75809
IUPAC Name 4-bromobutanoic acid
SMILES OC(=O)CCCBr

Specifications

Melting Point 31°C to 33°C
Boiling Point 129°C to 131°C (11 mmHg)
Flash Point 115°C (239°F)
Odor Stench
Quantity 10 g
UN Number UN3261
Beilstein 1743056
Sensitivity Light sensitive
Solubility Information Soluble in water and dilute alkali.
Formula Weight 167.01
Percent Purity 97%
Chemical Name or Material 4-Bromobutyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.